missing translation for 'onlineSavingsMsg'
Learn More

Potassium Carbonate, puriss p.a, ACS, ≥99.0% (T), Honeywell™ Fluka™

Product Code. 15625920 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
25 kg
4 x 2.5 kg
Packaging:
Poly bottle
Unit Size:
1kg
2.5kg
250g
25kg
500g
Pack of 4
This item is not returnable. View return policy

Product Code. 15625920

Brand: Honeywell-Fluka 60109500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, anhydrous,≥99.0% (T)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 584-08-7
Molecular Formula CK2O3
Molecular Weight (g/mol) 138.21
MDL Number MFCD00011382
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
Synonym potassium carbonate, dipotassium carbonate, potash, potassium carbonate, anhydrous, carbonic acid, dipotassium salt, carbonate of potash, pearl ash, salt of tartar, kaliumcarbonat, potassiumcarbonate
PubChem CID 11430
IUPAC Name dipotassium;carbonate
SMILES [K+].[K+].[O-]C([O-])=O

Specifications

Melting Point 891°C
Color Colorless or White
Quantity 500 g
Linear Formula K2CO3
UN Number NONH for all modes of transport
Beilstein 4267587
Formula Weight 138.21g/mol
Percent Purity ≥99.0% (T)
Packaging Poly bottle
Density 2.430g/cm3
Chemical Name or Material Potassium carbonate
Show More Show Less
compliance-icons
Product Identifier
  • Potassium carbonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Potassium carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.