missing translation for 'onlineSavingsMsg'
Learn More

Potassium carbonate, Purum p.a., anhydrous, ≥99.0% (T), Solstice

Product Code. 16693954
Change view
Click to view available options
20 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16693954 20 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16693954 Supplier Fluka Supplier No. 6011020KG
  • ONLINE EXCLUSIVE PRICE
    1280.00 €
    1201.65 € / 20kg
    Save 78.35 €
    6% Off
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 891°C
CAS 584-08-7
Color White
CAS Max % ≤ 100%
pH 11.6
Quantity 20 kg
Molecular Formula CK2O3
Linear Formula K2CO3
MDL Number MFCD00011382
Beilstein 4267587
InChI Key BWHMMNNQKKPAPP-UHFFFAOYSA-L
SMILES [K+].[K+].[O-]C([O-])=O
IUPAC Name dipotassium carbonate
Molecular Weight (g/mol) 138.21
Grade Reagent
Density 2.430 g/cm3 (19°C)
Chemical Name or Material Potassium carbonate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.