missing translation for 'onlineSavingsMsg'
Learn More

Potassium chlorate, 99+%

Product Code. 11488936
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11488936 250 g 250g
11478936 1000 g 1000g
11498936 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 11488936 Supplier Thermo Scientific Alfa Aesar Supplier No. A17075.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

CAS: 3811-04-9 | ClKO3 | 122.55 g/mol

Potassium chlorate is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium chlorate is used in textile printing and dyeing, as a disinfectant, in manufacturing of aniline black and other dyes. It can be utilized as a source of oxygen and also used in pyrotechnics and chemistry demonstrations. Potassium chlorate is useful for the flowering of dimocarpus longan and is a highly effective method for off-season fruit production. Potassium chlorate-osmium tetroxide oxidizing system has a useful application for the synthesis of alpha-dicarbonyl derivatives of acetylenic compounds and 2,3-dihydroxy-1,4-dione from a 2,5-dialkylfuran.

Solubility
Soluble in water, glycerol. Insoluble in alcohol.

Notes
Store away from incompatible materials. Store in a well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3811-04-9
Molecular Formula ClKO3
Molecular Weight (g/mol) 122.55
MDL Number MFCD00011361
InChI Key VKJKEPKFPUWCAS-UHFFFAOYSA-M
Synonym potassium chlorate, fekabit, potcrate, kaliumchlorat, anforstan, berthollet salt, salt of tarter, berthollet's salt, chlorate of potash, chloric acid, potassium salt
PubChem CID 6426889
IUPAC Name potassium;chlorate
SMILES [O-]Cl(=O)=O.[K+]

Specifications

Boiling Point 400°C (decomposition)
Melting Point 368°C
Quantity 250 g
Odor Odorless
Sensitivity Hygroscopic
Density 2.32
UN Number UN1485
Merck Index 14,7620
Solubility Information Soluble in water,glycerol. Insoluble in alcohol.
Formula Weight 122.55
Percent Purity ≥99%
Chemical Name or Material Potassium chlorate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium chlorate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Oxidising solid Category 1
Hazard Statement
  • H271-May cause fire or explosion; strong oxidiser.
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
Supplemental information
  • MIXTURE LIST-Contain: Potassium chlorate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.