missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, 3M aq. soln.

Product Code. 11335677
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11335677 250 mL 250mL
11345677 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11335677 Supplier Thermo Scientific Alfa Aesar Supplier No. 043398.AK
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For Refilling Calomel Electrodes | CAS: 7447-40-7 | ClK | 74.55 g/mol

For refilling calomel electrodesPotassium chloride, 3M aq. solution is used for volumetric analysis in laboratories.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7 , 7732-18-5
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium chloride
SMILES [Cl-].[K+]

Specifications

Color Colorless
Physical Form Liquid
Quantity 250 mL
Assay Percent Range 3M
Solubility Information Miscible with water.
Formula Weight 74.55
Concentration or Composition (by Analyte or Components) Potassium chloride: 23%; Water: 77%
Odor Odorless
Vapor Pressure 17mm Hg at 20°C
Chemical Name or Material Potassium chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.