missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, ≥99.0%, Reagent Grade, Honeywell™

Product Code. 15668030 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly bottle
Unit Size:
1kg
500g
5kg
This item is not returnable. View return policy

Product Code. 15668030

Brand: Honeywell Research Chemicals P4504500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
IUPAC Name potassium;chloride
SMILES [Cl-].[K+]

Specifications

Boiling Point 1413°C (1013hPa)
Melting Point 772°C
Color Colorless
pH 5 to 8 (at 20°C)
Physical Form Solid
Quantity 500 g
Linear Formula KCl
UN Number NONH for all modes of transport
Formula Weight 74.55g/mol
Percent Purity ≥99.0%
Odor Odorless
Packaging Poly bottle
Density 1.98g/mL at 25°C
Chemical Name or Material Potassium chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.