missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, ACS, 99.0-100.5%

Product Code. 11337337
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337337 5 kg 5kg
15490997 250 g 250g
11347337 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337337 Supplier Thermo Scientific Alfa Aesar Supplier No. 011595.A7
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy | CAS: 7447-40-7 | ClK | 74.55 g/mol

Potassium chloride is used to replenish electrolytes and restore water electrolyte balance in treating hypokalemia. It is also used in buffers, fertilizers and explosives and finds useful in medicine, scientific applications, and food processing. It plays an important role for the extraction and solubilization of proteins. It acts as a fire extinguishing agent in portable and wheeled fire extinguishers. It may be used as a softener unit in household water, which is an alternative to sodium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Boiling Point 1500°C (sublimation)
Melting Point 773°C
Physical Form Crystalline
Quantity 5 kg
Assay Percent Range 99.0 to 100.5%
Merck Index 14,7621
Solubility Information Soluble in water,glycerol and alkalies; Slightly soluble in alcohol and insoluble in ether.
IUPAC Name potassium;chloride
Formula Weight 74.55
Percent Purity 99.0 to 100.5%
Grade ACS
Refractive Index 1.49
Sensitivity Hygroscopic
Density 1.984 g/mL
Chemical Name or Material Potassium chloride, For analysis ACS
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.