missing translation for 'onlineSavingsMsg'
Learn More

Potassium Chloride, Certified AR for Analysis, Fisher Chemical

Product Code. 10684732 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1 kg
Packaging:
Plastic powder jar
Unit Size:
1kg
This item is not returnable. View return policy

Product Code. 10684732

Brand: Fisher Chemical P/4280/60

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Boiling Point 1420°C
Melting Point 770°C
CAS 7447-40-7
Color White
pH 6
Physical Form Solid
Quantity 1 kg
Molecular Formula ClK
MDL Number MFCD00011360
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.55
PubChem CID 4873
ChEBI CHEBI:32588
Formula Weight 74.54g/mol
Packaging Plastic powder jar
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.