missing translation for 'onlineSavingsMsg'
Learn More

Potassium chloride, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E508, 99 to 100.5% (AT), ≤0.0001% Al, Honeywell Fluka™
BIOPHARMA

Product Code. 15641170 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
2.5 kg
5 kg
25 kg
4 x 5 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
1kg
2.5kg
250g
25kg
500g
5kg
Pack of 4
Pack of 6
Product Code. Quantity unitSize
15641170 500 g 500g
15611170 250 g 250g
15691160 1 kg 1kg
15601170 2.5 kg 2.5kg
15651170 5 kg 5kg
15661170 6 x 1 kg Pack of 6
15671170 6 x 2.5 kg Pack of 6
15631170 4 x 5 kg Pack of 4
15621170 25 kg 25kg
9 options
This item is not returnable. View return policy

Product Code. 15641170

Brand: Honeywell-Fluka 12636500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E508, 99-100.5% (AT), ≤0.0001% Al

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-40-7
Molecular Formula ClK
Molecular Weight (g/mol) 74.55
MDL Number MFCD00011360
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Synonym potassium chloride, enseal, muriate of potash, klotrix, sylvite, slow-k, potassium chloride kcl, klor-con, chlorvescent, kalitabs
PubChem CID 4873
ChEBI CHEBI:32588
SMILES [Cl-].[K+]

Specifications

Quantity 500 g
Linear Formula KCl
UN Number NONH for all modes of transport
IUPAC Name potassium chloride
Formula Weight 74.55g/mol
Percent Purity 99 to 100.5% (AT)
Grade puriss.
Packaging Plastic Bottle
Density 1.98g/mL at 25°C
Chemical Name or Material Potassium Chloride, Meets analytical specification of Ph. Eur.BP, USP, FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.