missing translation for 'onlineSavingsMsg'
Learn More

SI Analytics™ Potassium Chloride Solution

Product Code. 10726911 Shop All SI Analytics Products
Click to view available options
Capacity (Metric):
1000 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10726911

Brand: SI Analytics™ 285138587

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

SI Analytics™ Potassium Chloride Solution is an aqueous electrolyte solution for reference electrodes.

  • Electrolyte solutions
  • Aqueous for reference electrodes
  • Electrolyte bridges
  • For storage
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Composition Potassium Chloride
Concentration 4.2 mol/L
Packaging DURAN™ Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.