Learn More
Potassium, chunks, in mineral oil, 98%
Description
Potassium forms a blue solution in tetrahydrofoliate which is used for the ? -monoalkylation of -lactones in the presence of 18-crown-6. It is used for generation of potassium ethoxide in solution, for use in the Birch reduction of an aromatic ring, for preparation of potassium amide,
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Potassium forms a blue solution in tetrahydrofoliate which is used for the ɑ -monoalkylation of -lactones in the presence of 18-crown-6. It is used for generation of potassium ethoxide in solution, for use in the Birch reduction of an aromatic ring, for preparation of potassium amide,
Solubility
Reacts with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Handle and store under inert gas. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Color | Gray-Silver |
| Melting Point | 63°C to 64°C |
| Boiling Point | 765°C to 770°C |
| Density | 0.860 g/mL |
| Quantity | 100 g |
| Odor | Odorless |
| Sensitivity | Air and moisture sensitive |
| Form | Solid |
| UN Number | UN2257 |
| Merck Index | 14,7604 |
| Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.