missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanate, 97%, pure

Product Code. 10720711 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
500g
This item is not returnable. View return policy

Product Code. 10720711

Brand: Thermo Scientific Acros 201110010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 590-28-3 | CKNO | 81.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-28-3
Molecular Formula CKNO
Molecular Weight (g/mol) 81.12
MDL Number MFCD00011411
InChI Key GKKCIDNWFBPDBW-UHFFFAOYSA-M
Synonym potassium cyanate, kaliumcyanat, kocn, aero cyanate, cyanic acid, potassium salt, alicyanate, bulpur, potassiumcyanate, weedanol cyanol, caswell no. 688
PubChem CID 11378442
ChEBI CHEBI:38904
IUPAC Name potassium;cyanate
SMILES [K+].[O-]C#N

Specifications

Melting Point 315.0°C
Color White
Physical Form Crystalline Powder and/or Chunks
Quantity 1 kg
Assay Percent Range 97%
Linear Formula KOCN
Fieser 01,1059
Merck Index 15, 7746
Solubility Information Solubility in water: 750g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 81.12
Percent Purity 97%
Grade Pure
Packaging Plastic bottle
Chemical Name or Material Potassium cyanate, Pure
Show More Show Less
compliance-icons
Product Identifier
  • Potassium cyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.