missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanide, 98+%, for analysis, Thermo Scientific Chemicals

Product Code. 10534274 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
2.5 kg
Unit Size:
100g
2.5kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10534274 100 g 100g
10495295 250 g 250g
10134260 500 g 500g
10247403 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10534274 Supplier Thermo Scientific Acros Supplier No. 196601000
  • ONLINE EXCLUSIVE PRICE
    52.15 €
    48.90 € / 100g
    Save 3.25 €
    6% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire: Des restrictions peuvent s'appliquer à l'achat de ce produit et nous pouvons être amenés à vous demander des documents supplémentaires.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.12
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym zyankali, cyanure de potassium french, kalium cyanid german, potassium cyanide k cn, unii-mqd255m2zo, potassium cyanide, caswell no. 688a, rcra waste number p098, cyanide of potassium
PubChem CID 9032

Spécification

Boiling Point 1625.0°C
Melting Point 634.0°C
Color Colorless
Physical Form Powder/Solid
Quantity 100 g
Assay Percent Range 97% min. (Argentometry)
Linear Formula CKN
Fieser 05,553; 07,299; 08,409; 10,324; 11,433; 13,87; 16,22
Merck Index 15, 7747
Solubility Information Solubility aq. soln.: 400 g/L (20°C). Other solubilities: ethanol 4.5 g/L (19.5°C)
SMILES [K+].[C-]#N
IUPAC Name potassium iminomethanide
Formula Weight 65.12
Percent Purity ≥97%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Potassium cyanide, p.a.
Afficher plus Afficher moins
compliance-icons compliance-icons compliance-icons compliance-icons
Identifiant produit
  • Potassium cyanide
Mention d'avertissement
  • Danger
Catégorie de danger
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1
Mention de danger
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H290-May be corrosive to metals.
  • EUH032-Contact with acids liberates very toxic gas.
Conseils de prudence
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.