missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanide, 97+%, Thermo Scientific Chemicals

Product Code. 11476790
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476790 500 g 500g
11466790 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11476790 Supplier Thermo Scientific Alfa Aesar Supplier No. L13273.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Potassium cyanide is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium cyanide is widely applied in organic synthesis for the synthesis of many organic compounds, for example in the synthesis of hydantoins, spiro heterocycles, nitriles, carboxylic acids, cyanohydrins and in von richter reaction. It is employed in gold mining, electroplating, and as a reagent in protein characterization.

Solubility
Highly soluble in water. Soluble in methanol, glycerol, and formamide. Slightly soluble in ethanol.

Notes
Deliquescent solid. Store in airtight containers. Incompatible with acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Boiling Point 1625°C
Melting Point 634°C
Quantity 500 g
Linear Formula KCN
UN Number UN1680
Beilstein 4652394
Merck Index 14,7626
Solubility Information Highly soluble in water. Soluble in methanol,glycerol,and formamide. Slightly soluble in ethanol.
Formula Weight 65.12
Percent Purity ≥97%
Sensitivity Hygroscopic
Density 1.52
Chemical Name or Material Potassium cyanide
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Substances and mixtures corrosive to metals Category 1
  • Specific target organ toxicity Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: Potassium cyanide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.