missing translation for 'onlineSavingsMsg'
Learn More

Potassium cyanide, ACS Reagent, ≥96.0%, Honeywell Fluka™

Product Code. 15651790 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Poly Bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15651790 500 g Poly Bottle 500g
15641790 25 g Poly Bottle 25g
15631790 2.5 kg Poly Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15651790 Supplier Honeywell-Fluka Supplier No. 207810500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

ACS Reagent,≥96.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 151-50-8
Molecular Formula CKN
Molecular Weight (g/mol) 65.116
MDL Number MFCD00011397
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
PubChem CID 9032
IUPAC Name potassium;cyanide
SMILES [C-]#N.[K+]

Specifications

Melting Point 634°C (literature)
Color White
Physical Form Solid
Quantity 500 g
Linear Formula KCN
UN Number UN1680
Beilstein 3593645
Solubility Information Soluble in water
Formula Weight 65.12g/mol
Percent Purity ≥96.0%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Granular Powder
Vapor Pressure 1.8 hPa at 634.5°C
Density 1.56g/cu.cm; Bulk Density: ca. 750kg/cu.m
Chemical Name or Material Potassium Cyanide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.