missing translation for 'onlineSavingsMsg'
Learn More

Potassium Cyanide, Extra Pure, SLR, Fisher Chemical

Product Code. 10599380 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10599380 250 g Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10599380 Supplier Fisher Chemical Supplier No. P/4560/50
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Specifications

Boiling Point 1625°C
Melting Point 634°C
CAS 151-50-8
Color White
pH 11 to 12
Physical Form Powder/Solid
Quantity 250 g
Molecular Formula CKN
MDL Number 11397
Synonym potassium cyanide, potassium cyanide k cn, cyanide of potassium, caswell no. 688a, zyankali, kalium cyanid german, unii-mqd255m2zo, rcra waste number p098, cyanure de potassium french
InChI Key NNFCIKHAZHQZJG-UHFFFAOYSA-N
SMILES [C-]#N.[K+]
IUPAC Name potassium;cyanide
Molecular Weight (g/mol) 65.116
PubChem CID 9032
Formula Weight 65.12g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Potassium cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.