missing translation for 'onlineSavingsMsg'
Learn More

Potassium dichromate, 99.5%, for analysis

Product Code. 10626972 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
500 g
2 kg
Packaging:
Plastic bottle
Unit Size:
2kg
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10626972 2 kg Plastic bottle 2kg
10092560 50 g Plastic bottle 50g
10563751 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10626972 Supplier Thermo Scientific Acros Supplier No. 196590020
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 7778-50-9 | Cr2K2O7 | 294.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-50-9
Molecular Formula Cr2K2O7
Molecular Weight (g/mol) 294.19
MDL Number MFCD00011367
InChI Key KMUONIBRACKNSN-UHFFFAOYSA-N
Synonym potassium dichromate, potassium bichromate, kaliumdichromat, iopezite, dipotassium bichromate, dipotassium dichromate, potassium dichromate vi, dichromic acid dipotassium salt, dipotassium dichromium heptaoxide, bichromate of potash
PubChem CID 24502
ChEBI CHEBI:53444
IUPAC Name dipotassium;oxido-(oxido(dioxo)chromio)oxy-dioxochromium
SMILES [O-][Cr](=O)(=O)O[Cr](=O)(=O)[O-].[K+].[K+]

Specifications

Boiling Point 500.0°C
Melting Point 398.0°C
Color Orange
Physical Form Fine Crystals or Crystalline Powder
Quantity 2 kg
Assay Percent Range 99.5% min. (Iodometry)
Linear Formula K2Cr2O7
Fieser 08,410; 09,383; 10,324; 12,405
Merck Index 15, 7748
Solubility Information Solubility in water: 125g/L (20°C). Other solubilities: 50 g/100 mL water (100°C), insoluble in alcohol
Formula Weight 294.19
Percent Purity 99.5%
Grade Analytical
Loss on Drying 0.05% max. (105°C)
Packaging Plastic bottle
Chemical Name or Material Potassium dichromate, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.