missing translation for 'onlineSavingsMsg'
Learn More

Potassium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10409380 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
3 kg
Unit Size:
1kg
250g
3kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10409380 3 kg 3kg
10284010 250 g 250g
10783611 500 g 500g
10793611 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10409380 Supplier Fisher Chemical Supplier No. P/4800/63
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potassium Dihydrogen Orthophosphate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 7778-77-0, White, Melting Point: 253 deg.C, Molecular Formula: H2KO4P, Molecular Weight: 136.084, Packaging: Plastic powder jar, pH: 4.5

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
Synonym potassium dihydrogen phosphate, monopotassium phosphate, potassium phosphate monobasic, potassium phosphate, monobasic, potassium acid phosphate, phosphoric acid, monopotassium salt, monobasic potassium phosphate, monopotassium monophosphate, monopotassium dihydrogen phosphate, monopotassium orthophosphate
PubChem CID 516951
ChEBI CHEBI:63036
SMILES [K+].OP(O)([O-])=O

Specifications

Melting Point 253°C
Color White
pH 4.5
Physical Form Solid
Quantity 3 kg
IUPAC Name potassium;dihydrogen phosphate
Formula Weight 136.09g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.