missing translation for 'onlineSavingsMsg'
Learn More

Potassium Disulfate, Puriss. p.a. ≥97.5% (T), Solstice

Product Code. 15605950 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15605950 1 kg Poly bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15605950 Supplier Honeywell-Fluka Supplier No. 602351KG
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss. p.a.,≥97.5% (T)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 325°C
CAS 7790-62-7
Color White
pH 1.0 to 2.0 (25.5g/L at 25°C)
Physical Form Solid
Quantity 1 kg
Molecular Formula K2O7S2
MDL Number MFCD00011385
UN Number UN3260
Synonym potassium pyrosulfate, dipotassium disulfate, potassium disulfate, unii-avj6zst7l6, avj6zst7l6, disulfuric acid dipotassium salt, dipotassium sulfonato sulfate, dipotassium disulphate, dipotassium pyrosulfate, kaliumpyrosulfat
InChI Key KAQHZJVQFBJKCK-UHFFFAOYSA-L
SMILES [O-]S(=O)(=O)OS(=O)(=O)[O-].[K+].[K+]
IUPAC Name dipotassium;sulfonato sulfate
Molecular Weight (g/mol) 254.31
PubChem CID 62681
Formula Weight 254.32g/mol
Percent Purity ≥97.5% (T)
Packaging Poly bottle
Density 2.28g/mL at 25°C
Chemical Name or Material Potassium disulfate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium disulphate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Dipotassium disulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.