missing translation for 'onlineSavingsMsg'
Learn More

Potassium disulfite, ≥98%, Honeywell™ Fluka™

Product Code. 15618010 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15618010 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15618010 Supplier Honeywell-Fluka Supplier No. P2522500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 16731-55-8
Quantity 500 g
Molecular Formula K2O5S2
Linear Formula K2S2O5
MDL Number MFCD00167605
UN Number NONH for all modes of transport
Synonym potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O
IUPAC Name dipotassium sulfinatosulfonate
Molecular Weight (g/mol) 222.31
PubChem CID 28019
Formula Weight 222.32g/mol
Percent Purity ≥98%
Packaging Poly Bottle
Density 2.3g/cm3
Chemical Name or Material Potassium disulfite
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.