missing translation for 'onlineSavingsMsg'
Learn More

Potassium Disulfite, puriss., Honeywell Fluka™

Product Code. p-7071785 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
Unit Size:
1kg
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15621160 1 kg 1kg
15641160 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15621160 Supplier Honeywell-Fluka Supplier No. 126191KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Puriss., meets analytical specification of Ph.Eur., BP, NF, FCC, E224, 95.0-101.0% (iodometric), powder, 51.8-57.6% sulfur dioxide

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16731-55-8
Molecular Formula K2O5S2
Molecular Weight (g/mol) 222.31
MDL Number MFCD00167605
InChI Key RWPGFSMJFRPDDP-UHFFFAOYSA-L
Synonym potassium metabisulfite, potassium disulfite, potassium pyrosulfite, dipotassium disulfite, dipotassium pyrosulfite, dipotassium metabisulfite, dipotassium disulphite, unii-65oe787q7w, ccris 1427, potassium disulfite k2s2o5
PubChem CID 28019
IUPAC Name dipotassium sulfinatosulfonate
SMILES [K+].[K+].[O-]S(=O)S([O-])(=O)=O

Specifications

Physical Form Powder
Quantity 1 kg
Linear Formula K2S2O5
UN Number NONH for all modes of transport
Formula Weight 222.32g/mol
Percent Purity 95 to 101% (iodometric)
Grade puriss.
Chemical Name or Material Potassium Disulfite, Meets analytical specification of Ph.Eur.BP, NF, FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.