missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium ferricyanide, 98%, pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
1kg
500g
5kg
Beskrivelse
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
Tekniske data
Tekniske data
| Color | Orange to Red |
| Physical Form | Crystalline Powder and/or Crystals |
| Quantity | 500 g |
| Assay Percent Range | 97.5% min. (Iodometry) |
| Linear Formula | K3Fe(CN)6 |
| Fieser | 01,929; 02,345; 04,406; 05,554; 06,480; 07,300; 08,410; 09,385; 13,255 |
| Merck Index | 15, 7751 |
| Solubility Information | Solubility in water: 464g/L (20°C). Other solubilities: slightly soluble in alcohol |
| IUPAC Name | tripotassium;iron(3+);hexacyanide |
| Formula Weight | 329.26 |
| Vis mere |
RUO – Research Use Only
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion