missing translation for 'onlineSavingsMsg'
Learn More

Potassium ferricyanide, 99+%, for analysis

Product Code. 10266370 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10266370 500 g Glass bottle 500g
10414583 100 g Glass bottle 100g
10390734 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10266370 Supplier Thermo Scientific Acros Supplier No. 223115000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13746-66-2 | C6FeK3N6 | 329.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Color Orange to Red
Physical Form Crystals and/or Chunks
Quantity 500 g
Assay Percent Range 99% min. (Iodometry)
Linear Formula K3Fe(CN)6
Fieser 01,929; 02,345; 04,406; 05,554; 06,480; 07,300; 08,410; 09,385; 13,255
Merck Index 15, 7751
Solubility Information Solubility in water: 464g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name tripotassium hexacyanoirontris(ylium)
Formula Weight 329.26
Percent Purity ≥99%
Grade Analytical
Packaging Glass bottle
Density 1.85
Chemical Name or Material Potassium ferricyanide, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.