missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferricyanide, BAKER ANALYZED™ ACS Reagent, J.T. Baker™

Product Code. 15558084 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
2.5kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15558084 2.5 kg Wide Mouth Amber Glass Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15558084 Supplier Avantor J.T.Baker Supplier No. 310405

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 13746-66-2
Physical Form Crystals
Quantity 2.5 kg
Molecular Formula C6FeK3N6
MDL Number MFCD00011392
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
IUPAC Name tripotassium;iron(3+);hexacyanide
Molecular Weight (g/mol) 329.25
PubChem CID 26250
Formula Weight 329.26
Grade Meets ACS reagent chemical requirements
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.85kg
Chemical Name or Material Potassium Ferricyanide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.