missing translation for 'onlineSavingsMsg'
Learn More

Potassium Ferricyanide, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15139414 Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15139414

Brand: Avantor J.T.Baker 310401

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 13746-66-2
Physical Form Crystals
Quantity 500 g
Molecular Formula C6FeK3N6
MDL Number MFCD00011392
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N
IUPAC Name tripotassium;iron(3+);hexacyanide
Molecular Weight (g/mol) 329.25
PubChem CID 26250
Formula Weight 329.26
Grade BAKER ANALYZED™ ACS Reagent
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 1.85kg
Chemical Name or Material Potassium Ferricyanide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.