missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Ferrocyanide, Trihydrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Shop All J.T.Baker Products
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
Specifications
Specifications
| Physical Form | Crystals |
| Quantity | 500 g |
| Molecular Formula | K4Fe(CN)6·3H2O |
| Formula Weight | 422.41 |
| Grade | BAKER ANALYZED™ ACS Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1L = 1.85kg |
| Chemical Name or Material | Potassium Ferrocyanide, Trihydrate |
Safety and Handling
Product Identifier
- Potassium hexacyanoferrate (II) trihydrate
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contains : Potassium hexacyanoferrate (II) trihydrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction