missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10136683 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10136683 1 kg Plastic bottle 1kg
10469910 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10136683 Supplier Thermo Scientific Acros Supplier No. 212600010
  • ONLINE EXCLUSIVE PRICE
    159.00 €
    154.00 € / 1kg
    Save 5.00 €
    3% Off
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride kf, potassium fluorure, potassium fluoride, anhydrous, potassiumfluoride, potassium fluoride on calcium fluoride, potassium fluorure french, unii-9082wg1g3f, potassium fluoride, ccris 1568, fluorure de potassium
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium fluoride
SMILES [F-].[K+]

Specifications

Boiling Point 1505.0°C
Melting Point 860.0°C
Color White
Physical Form Fine Crystalline Powder
Quantity 1 kg
Assay Percent Range 99+%
Linear Formula KF
Fieser 01,933; 02,346; 05,555; 06,481; 08,410; 10,325; 11,434; 12,406; 13,256; 15,167; 16,282
Merck Index 15, 7753
Solubility Information Solubility aq. soln.: 920 g/L (18°C). Other solubilities: soluble in hydrofluoric acid, insoluble in alcohol unless water is present
Formula Weight 58.09
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 2.48
Chemical Name or Material Potassium fluoride, p.a.
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.