missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Potassium fluoride, 99+%, for biochemistry, anhydrous

Product Code. 12695127 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12695127 100 g 100g
12605137 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12695127 Supplier Thermo Scientific Chemicals Supplier No. 447991000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

For Biochemistry

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-23-3
Molecular Formula FK
Molecular Weight (g/mol) 58.10
MDL Number MFCD00011398
InChI Key NROKBHXJSPEDAR-UHFFFAOYSA-M
Synonym potassium fluoride, potassiumfluoride, potassium fluoride kf, potassium fluorure, fluorure de potassium, potassium fluorure french, potassium fluoride on calcium fluoride, potassium fluoride, anhydrous, unii-9082wg1g3f, ccris 1568
PubChem CID 522689
ChEBI CHEBI:66872
IUPAC Name potassium;fluoride
SMILES [F-].[K+]

Specifications

Quantity 100 g
Assay Percent Range 99+%
Formula Weight 58.1
Chemical Name or Material Potassium fluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.