Learn More
Potassium fluoride, ACS, 99% min, Thermo Scientific Chemicals
Description
Potassium fluoride is used in metal finishing, batteries, coatings and photographic chemicals. It is utilized for the study of ion-specific swelling and de-swelling of ampholytic polymer gels as well as in the measurement of electronic polarizabilities of ions in polymers of alkali halides. It finds application in the electronic industry as a metal surface treatment product. It is used as a preservative, a food additive, a catalyzer and a water absorbing agent. In the Finkelstein reaction, it is actively involved in the conversion of chlorocarbons to fluorocarbons using polar solvents like dimethyl formamide and ethylene glycol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 1505°C |
| Melting Point | 859°C |
| Color | White |
| Physical Form | Powder |
| Quantity | 100 g |
| Assay Percent Range | 99% min |
| Linear Formula | KF |
| UN Number | UN1812 |
| Beilstein | 3902818 |
| Merck Index | 14,7632 |
| Show More |
Safety and Handling
- Potassium fluoride, ACS
- Danger
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Potassium fluoride
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.