Learn More
Potassium fluoride dihydrate, 98.5+%, Thermo Scientific Chemicals
Description
Potassium fluoride dihydrate is used as a stationary phase in gas chromatography. It is also used for the conversion of chlorocarbons into fluorocarbons through Finkelstein reaction. Further, it is used as a fluxing agent of silver brazing and metallurgical processing. It serves as an analytical reagent, complexing agent and masking agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 41°C |
| Physical Form | Crystalline |
| Quantity | 250 g |
| Odor | Odorless |
| Refractive Index | 1.352 |
| Sensitivity | Hygroscopic |
| Density | 2.5 g/mL |
| UN Number | UN1812 |
| Merck Index | 14,7632 |
| Solubility Information | Freely soluble in waterSoluble in water; Insoluble in ethanol. |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.