missing translation for 'onlineSavingsMsg'
Learn More

Potassium fluoride dihydrate, 98%, pure, Thermo Scientific Chemicals

Product Code. 15179233 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15179233 1 kg Plastic bottle 1kg
10713401 5 g Plastic bottle 5g
10533571 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15179233 Supplier Thermo Scientific Acros Supplier No. 206400010
  • ONLINE EXCLUSIVE PRICE
    230.00 €
    215.00 € / 1kg
    Save 15.00 €
    6% Off
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13455-21-5
Molecular Formula FH4KO2
Molecular Weight (g/mol) 94.13
MDL Number MFCD00149929
InChI Key MZFXHSQFQLKKFD-UHFFFAOYSA-M
Synonym potassium fluoride dihydrate, unii-95r1d12heh, potassium dihydrate fluoride, potassium fluoride dihydrate, 98.5+%, potassium fluoride dihydrate, puratronic, potassium fluoride, dihydrate, potassium fluoride kf , dihydrate, acmc-1byyf, ksc497c4l, potassiumfluoridedihydrate
PubChem CID 23708904
SMILES O.O.[F-].[K+]

Specifications

Color Colorless to White
Physical Form Crystalline Powder and/or Crystals
Quantity 1 kg
Packaging Plastic bottle
Assay Percent Range 98%
Density 2.45
Linear Formula KF·2H2O
Fieser 01,933; 02,346; 05,555; 06,481; 08,410; 16,282
IUPAC Name potassium;fluoride;dihydrate
Formula Weight 94.13
Percent Purity 19.65 to 20.70% (F)
Grade Pure
Chemical Name or Material Potassium fluoride dihydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.