missing translation for 'onlineSavingsMsg'
Learn More

Potassium Formate, Purum p.a., ≥99.0% (NT), Honeywell™

Product Code. 15665950 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 g
Packaging:
Poly bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 15665950

Brand: Honeywell Research Chemicals 60243500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.0% (NT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Specifications

Boiling Point >167°C
Melting Point 165°C to 168°C
CAS 590-29-4
Color White
Physical Form Crystalline
Quantity 500 g
Molecular Formula CHKO2
Linear Formula HCOOK
MDL Number MFCD00013100
UN Number NONH for all modes of transport
Beilstein 3623272
Synonym potassium formate, formic acid potassium salt, formic acid, potassium salt, formic acid, k salt, hcook, mravencan draselny czech, hco2k, formic acid, potassium salt 1:1, potassium formira, 64-18-6 parent
InChI Key WFIZEGIEIOHZCP-UHFFFAOYSA-M
SMILES C(=O)[O-].[K+]
IUPAC Name potassium;formate
Molecular Weight (g/mol) 84.115
PubChem CID 2735122
ChEBI CHEBI:63316
Formula Weight 84.12g/mol
Percent Purity ≥99.0% (NT)
Odor Odorless
Packaging Poly bottle
Chemical Name or Material Potassium formate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.