missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexachloropalladate(IV), 98%, BLD Pharm™

Product Code. 30802093
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
5 g
25 g
Unit Size:
100mg
1g
250mg
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30802093 100 mg 100mg
30790236 250 mg 250mg
30799505 5 g 5g
30786814 1 g 1g
30800854 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 30802093 Supplier BLD Pharm Supplier No. BD148792100mg
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 16919-73-6
Quantity 100 mg
Molecular Formula Cl6K2Pd
MDL Number MFCD00043081
SMILES Cl[Pd-2](Cl)(Cl)(Cl)(Cl)Cl.[K+].[K+]
Molecular Weight (g/mol) 397.33
Percent Purity 98%
Chemical Name or Material Potassium hexachloropalladate(IV)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.