missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanocobaltate(III), 95%

Product Code. 10480861 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10480861

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13963-58-1 | C6CoK3N6 | 332.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13963-58-1
Molecular Formula C6CoK3N6
Molecular Weight (g/mol) 332.34
MDL Number MFCD00011391

Specifications

Color Brown to Yellow
Physical Form Crystalline Solid
Quantity 500 g
Assay Percent Range 95%
Linear Formula K3[Co(CN)6]
Merck Index 15, 7758
Solubility Information Solubility in water: freely soluble. Other solubilities: insoluble in alcohol
Formula Weight 332.34
Percent Purity 95%
Packaging Glass bottle
Specific Gravity 1.87
Density 1.8700g/mL
Chemical Name or Material Potassium hexacyanocobaltate(III), 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hexacyanocobaltate(III)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.