missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanocobaltate(III), ≥97.0%, Honeywell Fluka™

Product Code. 15601230 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
20 kg
Unit Size:
1kg
20kg
250g
This item is not returnable. View return policy

Product Code. 15601230

Brand: Honeywell-Fluka 1290220KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

≥97.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13963-58-1
MDL Number MFCD00011391
Synonym Tripotassium hexacyanocobaltate(III)

Specifications

CAS Max % 100.0000%
Quantity 20 kg
Linear Formula K3Co(CN)6
UN Number NONH for all modes of transport
Formula Weight 332.33
Percent Purity >97%
Packaging Paper / Poly Bag
Density 1.878 g/mL at 25 °C(lit.)
Chemical Name or Material Potassium hexacyanocobaltate(III), ≥97.0%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.