missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexacyanoferrate(II), Extra Pure, SLR, Fisher Chemical

Product Code. 10214160 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Plastic powder jar
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10214160 500 g Plastic powder jar 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10214160 Supplier Fisher Chemical Supplier No. P/4920/53
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 70°C
CAS 14459-95-1
Color Yellow
pH 9.5
Physical Form Powder/Solid
Quantity 500 g
Molecular Formula C6H6FeK4N6O3
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
Molecular Weight (g/mol) 422.39
Formula Weight 422.4g/mol
Packaging Plastic powder jar
Show More Show Less
Product Identifier
  • Potassium hexacyanoferrate (II)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contain: Potassium ferrocyanide trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.