missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hexacyanoferrate(II) Trihydrate, 98.5 to 102.0%, ACS Reagent, Honeywell™ Fluka™

Product Code. 15698010 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Poly bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15698010 5 g Glass bottle 5g
15678010 100 g Poly bottle 100g
15688010 500 g Poly bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15698010 Supplier Honeywell-Fluka Supplier No. P32895G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent, 98.5-102.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 70°C
Color Yellow
Quantity 5 g
Linear Formula K4Fe(CN)6 · 3H2O
UN Number NONH for all modes of transport
Formula Weight 422.39g/mol
Percent Purity 98.5 to 102.0%
Odor Odorless
Packaging Glass bottle
Appearance Crystalline
Density 1.85g/cu.cm at 20°C
Chemical Name or Material Potassium hexacyanoferrate(II) trihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.