missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanoferrate(II) trihydrate, 98+%

Product Code. 11442378
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
This item is not returnable. View return policy

Product Code. 11442378

Brand: Thermo Scientific Alfa Aesar A15736.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14459-95-1 | C6H6FeK4N6O3 | 422.39 g/mol

Potassium hexacyanoferrate(II) trihydrate is widely used in anticaking agent for both road and table salt. It is involved in the purification of tin and separation of copper from molybdenum ores. It plays a vital role in the in production of wine and citric acid. In analytical chemistry, it is used to determine the concentration of potassium permanganate. It is in combination with potassium ferricyanide and phosphate buffered solution acts as buffer for beta-galatosidase. Further, it is used as fertilizers for plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hexacyanoferrate(II) trihydrate is widely used in anticaking agent for both road and table salt. It is involved in the purification of tin and separation of copper from molybdenum ores. It plays a vital role in the in production of wine and citric acid. In analytical chemistry, it is used to determine the concentration of potassium permanganate. It is in combination with potassium ferricyanide and phosphate buffered solution acts as buffer for beta-galatosidase. Further, it is used as fertilizers for plants.

Solubility
Soluble in water. Insoluble in ethanol and ether.

Notes
Begins to lose water at 600C, anhydrous at 1000C. Incompatible with oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14459-95-1
Molecular Formula C6H6FeK4N6O3
Molecular Weight (g/mol) 422.39
MDL Number MFCD00167023
InChI Key NLYKFUFIUWPPIZ-UHFFFAOYSA-N
Synonym Potassium ferrocyanide trihydrate
IUPAC Name tetrapotassium hexacyanoironbis(ylium) trihydrate
SMILES O.O.O.[K+].[K+].[K+].[K+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point 69°C to 71°C
pH 9.5
Quantity 100 g
Merck Index 14,7631
Solubility Information Soluble in water. Insoluble in ethanol and ether.
Formula Weight 422.39 (368.36 Anhydrous)
Percent Purity ≥98%
Odor Odorless
Density 1.85
Chemical Name or Material Potassium hexacyanoferrate(II) trihydrate
compliance-icons
Product Identifier
  • Potassium hexacyanoferrate(II) trihydrate
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : potassium ferrocyanate trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.