missing translation for 'onlineSavingsMsg'
Learn More

Potassium hexacyanoferrate(III), 98+%

Product Code. 11458786
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11458786 250 g 250g
11448786 1000 g 1000g
11468786 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11458786 Supplier Thermo Scientific Alfa Aesar Supplier No. A16946.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 13746-66-2 | C6FeK3N6 | 329.25 g/mol

Potassium hexacyanoferrate(III) is used in reading palimpsests and old manuscripts, in blueprint drawing and in photography (Cyanotype process). It finds application to harden iron and steel, in electroplating, dyeing wool and as a laboratory reagent. It is a mild oxidizing agent used in organic synthesis. It is associated with potassium hydroxide solution used to formulate Murakami’s etchant, which is useful for metallographers to provide contrast between binder and carbide phases in cemented carbides. Further, it is used in many amperometric biosensors as an electron transfer agent replacing an enzyme's natural electron transfer agent such as oxygen with the enzyme glucose oxidase. It is an active ingredient in blood glucose meters used by diabetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hexacyanoferrate(III) is used in reading palimpsests and old manuscripts, in blueprint drawing and in photography (Cyanotype process). It finds application to harden iron and steel, in electroplating, dyeing wool and as a laboratory reagent. It is a mild oxidizing agent used in organic synthesis. It is associated with potassium hydroxide solution used to formulate Murakami’s etchant, which is useful for metallographers to provide contrast between binder and carbide phases in cemented carbides. Further, it is used in many amperometric biosensors as an electron transfer agent replacing an enzyme′s natural electron transfer agent such as oxygen with the enzyme glucose oxidase. It is an active ingredient in blood glucose meters used by diabetics.

Solubility
Soluble in water and acid. Slightly soluble in alcohol.

Notes
Light sensitive. Incompatible with strong acids, strong oxidizing agents, ammonia, hydrochloric acid and cyanides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13746-66-2
Molecular Formula C6FeK3N6
Molecular Weight (g/mol) 329.25
MDL Number MFCD00011392
InChI Key QMTKJUMXUDIUAQ-UHFFFAOYSA-N
Synonym potassium ferricyanide, red prussiate, potassium ferricyanate, tripotassium hexacyanoferrate, potassium cyanoferrate, iron potassium cyanide, tripotassium ferricyanide, tripotassium iron hexacyanide, tripotassium ferric hexacyanide, ccris 5559
PubChem CID 26250
SMILES [K+].[K+].[K+].N#C[Fe+3](C#N)(C#N)(C#N)(C#N)C#N

Specifications

Melting Point (decomposition)
Quantity 250 g
Linear Formula K3Fe(CN)6
Merck Index 14,7630
Solubility Information Soluble in water and acid. Slightly soluble in alcohol.
IUPAC Name tripotassium;iron(3+);hexacyanide
Formula Weight 329.26
Percent Purity ≥98%
Sensitivity Light sensitive
Density 1.89
Chemical Name or Material Potassium hexacyanoferrate(III)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.