missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium hexanitroiridate(III), Ir 32.3% min
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Descrizione
Potassium hexanitroiridate(III) is most frequently used in industrial and chemical applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Physical Form | Micro-crystals |
| Quantity | 5 g |
| Solubility Information | It is slightly soluble in water. It is soluble in organic or non-aqueous solvent. |
| IUPAC Name | tripotassium hexanitrite iridiumtriuide |
| Formula Weight | 585.54 |
| Concentration or Composition (by Analyte or Components) | Ir 32.3% min. |
| Chemical Name or Material | Potassium hexanitroiridate(III) |
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto