missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Difluoride, purum p.a, ≥99.0% (F), Honeywell™

Product Code. 15695950 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15695950 1 kg Poly bottle 1kg
15605960 250 g Poly bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15695950 Supplier Honeywell Research Chemicals Supplier No. 603441KG

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Purum p.a.,≥99.0% (F)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-29-9
MDL Number MFCD00011403

Specifications

Boiling Point >180°C
Melting Point 239°C
pH Acidic
Physical Form Solid
Quantity 1 kg
Linear Formula KHF2
UN Number UN1811
Solubility Information 392g/L
Formula Weight 78.1g/mol
Percent Purity ≥99.0% (F)
Packaging Poly bottle
Density 2.37g/cm3
Chemical Name or Material Potassium hydrogen difluoride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.