missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen fluoride, 98%, Thermo Scientific Chemicals

Product Code. 11444020
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444020 100 g 100g
11419963 500 g 500g
11429963 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11444020 Supplier Thermo Scientific Alfa Aesar Supplier No. A13031.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Potassium hydrogen fluoride is used as an abrasive, as a pretreatment of metals before they are further processed, as a wood preservative and as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen fluoride is used as an abrasive, as a pretreatment of metals before they are further processed, as a wood preservative and as a chemical intermediate.

Solubility
Soluble in water (392 g/L). Also soluble in ethanol.

Notes
It is hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-29-9
Molecular Formula KHF2
MDL Number MFCD00011403
Synonym Potassium bifluoride

Specifications

Melting Point 239°C (decomposition)
pH 1
Quantity 100 g
UN Number UN1811
Merck Index 14,7610
Solubility Information Soluble in water (392g/L). Also soluble in ethanol.
Formula Weight 78.1
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 2.37
Chemical Name or Material Potassium hydrogen fluoride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Potassium hydrogen fluoride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Potassium hydrogen fluoride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.