missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Fluoride, TRC

Product Code. 30425203
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425203 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30425203 Supplier Toronto Research Chemicals Supplier No. TRCP99331350G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Hydrogen Fluoride
CAS 7789-29-9
Quantity 50 g
Molecular Formula KHF2
Synonym Potassium fluoride hydrofluoride
InChI Formula InChI=1S/2FH.K/h2*1H;/q;;+1/p-1
SMILES F.[F-].[K+]
IUPAC Name potassium;fluoride;hydrofluoride
Molecular Weight (g/mol) 78.1
Formula Weight 77.9683
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.