missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phthalate, 99.99%, acidimetric standard

Product Code. 10428330 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10428330 500 g Plastic Bottle 500g
10418330 100 g Plastic Bottle 100g
11915051 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10428330 Supplier Thermo Scientific Acros Supplier No. 177125000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Melting Point 295.0°C to 300.0°C
Color White
pH 3.8 to 4 (5% soln. at 20°C)
Infrared Spectrum Authentic
Assay Percent Range 99.99%
Packaging Plastic Bottle
Molecular Formula C8H5KO4
Linear Formula 2-(HO2C)C6H4CO2K
Quantity 500 g
Beilstein 09, 791
Merck Index 15, 7733
Molecular Weight (g/mol) 204.222
Formula Weight 204.22
Percent Purity 99.95 to 100.05%
Grade Trace Metal Basis
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Potassium Hydrogen Phthalate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.