missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen phthalate, 99%

Product Code. 11445343
Change view
Click to view available options
Quantity:
1000 g
5000 g
Unit Size:
1000g
5000g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11445343 5000 g 5000g
11425343 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11445343 Supplier Thermo Scientific Alfa Aesar Supplier No. A12895.0I
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol

Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters and for the preparation of volumetric alkali solutions. It acts as buffer in pH determination. It finds application as a useful standard for total organic carbon testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium hydrogen phthalate is used as a primary standard for acid- base titrations as well as for calibrating pH meters and for the preparation of volumetric alkali solutions. It acts as buffer in pH determination. It finds application as a useful standard for total organic carbon testing.

Solubility
Soluble in water. Slightly soluble in alcohol.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
MDL Number MFCD00013070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Melting Point 297°C (decomposition)
Density 1.636
Odor Odorless
Quantity 5000 g
Beilstein 3637128
Merck Index 14,7612
Solubility Information Soluble in water. Slightly soluble in alcohol.
Formula Weight 204.23
Percent Purity 99%
Chemical Name or Material Potassium hydrogen phthalate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.