missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Phthalate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10040340 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Unit Size:
100g
1kg
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10040340 250 g 250g
10254300 100 g 100g
10437460 500 g 500g
10151960 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10040340 Supplier Fisher Chemical Supplier No. P/5320/50
  • ONLINE EXCLUSIVE PRICE
    95.23 €
    75.50 € / 250g
    Save 19.73 €
    20% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potassium Hydrogen Phthalate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 877-24-7, White, Melting Point: 295 deg.C, Molecular Formula: C8H5KO4, Molecular Weight: 204.222, Packaging: Plastic powder jar, pH: 3.8 to 4.0 | CAS: 877-24-7 | C8H5KO4 | 204.222 g/mol

Potassium Hydrogen Phthalate, Certified AR for Analysis, Fisher Chemical, Quantity: 500g, CAS: 877-24-7, White, Melting Point: 295 deg.C, Molecular Formula: C8H5KO4, Molecular Weight: 204.222, Packaging: Plastic powder jar, pH: 3.8 to 4.0

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 877-24-7
Molecular Formula C8H5KO4
Molecular Weight (g/mol) 204.222
MDL Number 13070
InChI Key IWZKICVEHNUQTL-UHFFFAOYSA-M
Synonym potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
PubChem CID 23676735
IUPAC Name potassium;2-carboxybenzoate
SMILES C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]

Specifications

Melting Point 295°C
Color White
pH 3.8 to 4.0
Physical Form Solid
Quantity 250 g
Formula Weight 204.22g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.