missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Sulfate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10408420 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10408420 250 g 250g
10408610 500 g 500g
10418610 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10408420 Supplier Fisher Chemical Supplier No. P/5440/50
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium Hydrogen Sulfate, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
SMILES [K+].OS([O-])(=O)=O

Specifica

Melting Point 214°C
Color White
pH 1
Physical Form Solid
Quantity 250 g
IUPAC Name potassium hydrogen sulfate
Formula Weight 136.16g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Identificativo prodotto
  • Potassium bisulfate
Avvertenza
  • Danger
Categoria di pericolo
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Consigli di prudenza
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.