missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Sulfate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10418610 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
500 g
1 kg
Unit Size:
1kg
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10418610 1 kg 1kg
10408420 250 g 250g
10408610 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10418610 Supplier Fisher Chemical Supplier No. P/5440/60
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potassium Hydrogen Sulfate, Certified AR for Analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-93-7
Molecular Formula HKO4S
Molecular Weight (g/mol) 136.16
MDL Number MFCD00011404
InChI Key CHKVPAROMQMJNQ-UHFFFAOYSA-M
Synonym potassium bisulfate, potassium hydrogen sulfate, potassium bisulphate, sulfuric acid, monopotassium salt, potassium acid sulfate, sal enixum, potassium hydrogensulphate, monopotassium sulfate, acid potassium sulfate, caswell no. 682c
PubChem CID 516920
SMILES [K+].OS([O-])(=O)=O

Specifications

Melting Point 214°C
Color White
pH 1
Physical Form Solid
Quantity 1 kg
IUPAC Name potassium;hydrogen sulfate
Formula Weight 136.16g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Potassium bisulfate
Signal Word
  • Danger
Catégorie de danger
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Mention de danger
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H314-Causes severe skin burns and eye damage.
Conseils de prudence
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Informations supplémentaires
  • MIXTURE LIST-Contain: Potassium hydrogen sulfate
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.