missing translation for 'onlineSavingsMsg'
Learn More

Potassium hydrogen sulfate, fused, ACS, mixture of potassium pyrosulfate, K{2}S{2}O{7}, and potassium bisulfate, KHSO{4}

Product Code. 11391339
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11391339 100 g 100g
11301349 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11391339 Supplier Thermo Scientific Alfa Aesar Supplier No. 013800.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potassium hydrogen sulfate is used in the conversion of tartrates to bitartrates in wine. It is also used as a disintegrating agent in analytical chemistry. Further, it is used to make fertilizers and in the preparation of potassium persulfate. In addition to this, it is used as a catalyst in organic preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00066385

Specifications

Melting Point 214°C
Color White
Physical Form Fused Powder
Quantity 100 g
Solubility Information Soluble in water,isopropyl alcohol and acetone.
Formula Weight 136.17
Grade ACS
Odor Odorless
Sensitivity Hygroscopic
Appearance White fused powder
Chemical Name or Material Potassium hydrogen sulfate, fused
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.