missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Hydrogen Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | Potassium Hydrogen Sulfate |
| CAS | 7646-93-7 |
| Quantity | 10 g |
| Molecular Formula | HKO4S |
| Synonym | Hydrogen Potassium Sulfate, Monopotassium Sulfate, Potassium Acid Sulfate, Potassium Bisulfate, Potassium bisulphate, Potassium Hydrogen Sulfate, Potassium Sulfate (KHSO4), Sal Enixum |
| InChI Formula | InChI=1S/K.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+1;/p-1 |
| SMILES | [K+].OS(=O)(=O)[O-] |
| IUPAC Name | potassium;hydrogen sulfate |
| Molecular Weight (g/mol) | 136.16 |
| Formula Weight | 135.9233 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?