missing translation for 'onlineSavingsMsg'
Learn More

Potassium Hydrogen Sulfate, TRC

Product Code. 30419716
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30419716 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30419716

Brand: Toronto Research Chemicals TRCP6990801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Hydrogen Sulfate
CAS 7646-93-7
Molecular Formula HKO4S
Quantity 1 g
Synonym Hydrogen Potassium Sulfate, Monopotassium Sulfate, Potassium Acid Sulfate, Potassium Bisulfate, Potassium bisulphate, Potassium Hydrogen Sulfate, Potassium Sulfate (KHSO4), Sal Enixum
InChI Formula InChI=1S/K.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+1;/p-1
SMILES [K+].OS(=O)(=O)[O-]
IUPAC Name potassium;hydrogen sulfate
Molecular Weight (g/mol) 136.16
Formula Weight 135.9233
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.